Gene Transcripts

Gene Symbol
Rgs5
Entrez Gene ID
19737
Gene Name
regulator of G-protein signaling 5
Gene Aliases
1110070A02Rik
Chromosome Location
Chr. 1: 169655501 - 169693526 on Build GRCm38
UniGene ID
Mm.20954
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009063.3 NP_033089.2 5 991
GenBank mRNA AK028765.1 - 994
AK030300.1 - 993
AK054098.1 - 5 992
BC037683.1 - 5 977 3962083

View Full Product Details