Gene Transcripts

Gene Symbol
Selenbp1
Entrez Gene ID
20341
Gene Name
selenium binding protein 1
Gene Aliases
Lp56, Lpsb, SBP56
Chromosome Location
Chr. 3: 94933083 - 94944758 on Build GRCm38
UniGene ID
Mm.196558
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009150.3 NP_033176.2 12 1345
GenBank mRNA AK002447.1 - 12 1361
BC011202.1 - 12 1345 4209393
M32032.1 - 1328

View Full Product Details