Gene Transcripts

Gene Symbol
Sgca
Entrez Gene ID
20391
Gene Name
sarcoglycan, alpha (dystrophin-associated glycoprotein)
Gene Aliases
50DAG, Asg
Chromosome Location
Chr. 11: 94962777 - 94976327 on Build GRCm38
UniGene ID
Mm.18709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009161.4 NP_033187.1 8, 9 1213
GenBank mRNA AB024920.1 - 8, 9 1213
AF019564.1 - 7, 8 960
AK009015.1 - 7, 8 971
AK075915.1 - 7, 8 985
BC060287.1 - 7, 8 965

View Full Product Details