Gene Transcripts
- Gene Symbol
- EPHA1
- Entrez Gene ID
- 2041
- Gene Name
- EPH receptor A1
- Gene Aliases
- EPH, EPHT, EPHT1
- Chromosome Location
- Chr. 7: 143390813 - 143408892 on Build GRCh38
- UniGene ID
- Hs.89839
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005232.4 |
NP_005223.4
|
1, 2 |
173 |
|
XM_006715880.3 |
XP_006715943.1
|
1, 2 |
146 |
|
GenBank mRNA |
AK290351.1 |
- |
1, 2 |
137 |
|
AK313263.1 |
- |
1, 2 |
146 |
|
AU131228.1 |
- |
1, 2 |
132 |
|
BC130291.1 |
- |
1, 2 |
157 |
40146322 |
BC144156.1 |
- |
1, 2 |
157 |
|
EU826604.1 |
- |
1, 2 |
86 |
|
EU826605.1 |
- |
1, 2 |
86 |
|
M18391.1 |
- |
1, 2 |
179 |
|
View Full Product Details