Gene Transcripts

Gene Symbol
Tectb
Entrez Gene ID
21684
Gene Name
tectorin beta
Gene Aliases
Tctnb
Chromosome Location
Chr. 19: 55180733 - 55196313 on Build GRCm38
UniGene ID
Mm.42139
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009348.3 NP_033374.2 2 334
GenBank mRNA BC138888.1 - 2 263
BC138889.1 - 2 263 8861911
X99806.2 - 2 334

View Full Product Details