Gene Transcripts
- Gene Symbol
- Sumo1
- Entrez Gene ID
- 22218
- Gene Name
- SMT3 suppressor of mif two 3 homolog 1 (yeast)
- Gene Aliases
- GMP1, PIC1, SENTRIN, SMT3, SMT3H3, SMTP3, SUMO-1, Smt3C, Ubl1
- Chromosome Location
- Chr. 1: 59639434 - 59670834 on Build GRCm38
- UniGene ID
- Mm.362118
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009460.2 |
NP_033486.1
|
2, 3 |
238 |
|
GenBank mRNA |
AF033353.1 |
- |
2, 3 |
213 |
|
AK002536.1 |
- |
2, 3 |
208 |
|
AK011074.1 |
- |
2, 3 |
207 |
|
AK089081.1 |
- |
2, 3 |
187 |
|
AK159366.1 |
- |
2, 3 |
209 |
|
AK220052.1 |
- |
1, 2 |
102 |
|
BC082566.1 |
- |
2, 3 |
213 |
30602751 |
BC083158.1 |
- |
2, 3 |
211 |
6478032 |
BY242724.1 |
- |
2, 3 |
239 |
|
View Full Product Details