Gene Transcripts

Gene Symbol
Nr1h2
Entrez Gene ID
22260
Gene Name
nuclear receptor subfamily 1, group H, member 2
Gene Aliases
AI194859, LXR, LXRB, LXRbeta, NER1, OR-1, RIP15, UR, Unr, Unr2
Chromosome Location
Chr. 7: 44549616 - 44553969 on Build GRCm38
UniGene ID
Mm.968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285517.1 NP_001272446.1 6 772
NM_001285518.1 NP_001272447.1 6 766
NM_001285519.1 NP_001272448.1 6 763
NM_009473.3 NP_033499.1 6 779
GenBank mRNA AK054508.1 - 6 750
AK077620.1 - 6 745
AK134489.1 - 6 728
AK156777.1 - 6 747
BC066025.1 - 6 730 6829527
BC106148.1 - 5 499
BQ573162.1 - 6 749
EU869275.2 - 3 758
U09419.1 - 5 597

View Full Product Details