Gene Transcripts

Gene Symbol
Wee1
Entrez Gene ID
22390
Gene Name
WEE 1 homolog 1 (S. pombe)
Gene Aliases
Wee1A
Chromosome Location
Chr. 7: 110121700 - 110143299 on Build GRCm38
UniGene ID
Mm.287173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009516.3 NP_033542.2 11 2275
GenBank mRNA AK011212.1 - 11 2051
AK039572.1 - 11 1591
BC006852.1 - 11 2294 3156953

View Full Product Details