Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009576.2 | NP_033602.2 | 3 | 917 | |
GenBank mRNA | AK028305.1 | - | 1 | 720 | |
AK035908.1 | - | 1 | 721 | ||
AK046913.1 | - | 3 | 918 | ||
AK163477.1 | - | 3 | 1991 | ||
D78174.1 | - | 1 | 1170 |