Gene Transcripts

Gene Symbol
BARX1
Entrez Gene ID
56033
Gene Name
BARX homeobox 1
Gene Aliases
Chromosome Location
Chr. 9: 93951627 - 93955326 on Build GRCh38
UniGene ID
Hs.164960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021570.3 NP_067545.3 4 1106
GenBank mRNA BC009458.1 - 3 953
BC064363.1 - 4 1151 5555949

View Full Product Details