Gene Transcripts

Gene Symbol
Edn1
Entrez Gene ID
13614
Gene Name
endothelin 1
Gene Aliases
ET-1, preproET
Chromosome Location
Chr. 13: 42301270 - 42307989 on Build GRCm38
UniGene ID
Mm.14543
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010104.3 NP_034234.1 2 816
GenBank mRNA AB081657.1 - 2 202
AK040778.1 - 2 474
AK152647.1 - 2 467
AK153437.1 - 2 471
BC029547.1 - 2 610 5059522
D43775.1 - 2 527
U35233.1 - 2 221

View Full Product Details