Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010111.5 | NP_034241.2 | 2 | 315 | |
GenBank mRNA | AK132512.1 | - | 2 | 782 | |
AK132845.1 | - | 2 | 608 | ||
AK134108.1 | - | 2 | 782 | ||
AK142203.1 | - | 2 | 785 | ||
AK142283.1 | - | 2 | 785 | ||
AK143059.1 | - | 2 | 782 | ||
BC057009.1 | - | 2 | 603 | 6827408 | |
CT010381.1 | - | 2 | 156 | ||
L38847.1 | - | 2 | 316 | ||
U16819.1 | - | 2 | 184 | ||
U30244.1 | - | 2 | 153 |