Gene Transcripts

Gene Symbol
En1
Entrez Gene ID
13798
Gene Name
engrailed 1
Gene Aliases
En-1, Mo-en.1
Chromosome Location
Chr. 1: 120602391 - 120607991 on Build GRCm38
UniGene ID
Mm.2657
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010133.2 NP_034263.2 2 1706
GenBank mRNA AK140408.1 - 2 1569
L12703.1 - 2 1512

View Full Product Details