Gene Transcripts

Gene Symbol
Dtna
Entrez Gene ID
13527
Gene Name
dystrobrevin alpha
Gene Aliases
2210407P21Rik, DTN-A, Dtn, Gm19389, a-DB-1, adbn
Chromosome Location
Chr. 18: 23310020 - 23659719 on Build GRCm38
UniGene ID
Mm.484047
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285807.1 NP_001272736.1 8 1125
NM_001285808.1 NP_001272737.1 6 781
NM_001285810.1 NP_001272739.1 6 781
NM_001285811.1 NP_001272740.1 7 903
NM_001285813.1 NP_001272742.1 7 938
NM_001285817.1 NP_001272746.1 6 781
NM_010087.4 NP_034217.2 7 903
NM_207650.4 NP_997533.1 7 903
GenBank mRNA AF143542.1 - 7 802
AF143543.1 - 7 839
AF143544.1 - 7 748
AK003829.2 - 7 823
AK019477.1 - 8 1045
AK131649.1 - 6 674
AK147279.1 - 7 849
AK147359.1 - 6 738
BC040364.1 - 7 870 4505583
BC050765.1 - 6 689 6533769
X95226.1 - 6 603
X95227.1 - 7 733

View Full Product Details