Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077353.2 | NP_001070821.1 | 8 | 870 | |
NM_001288617.1 | NP_001275546.1 | 8 | 915 | ||
NM_010356.4 | NP_034486.2 | 7 | 775 | ||
GenBank mRNA | AK002478.1 | - | 6 | 723 | |
AK014076.1 | - | 7 | 748 | ||
AK149543.1 | - | 7 | 740 | ||
BC147272.1 | - | 7 | 738 | 9056327 | |
BC147273.1 | - | 7 | 738 | 9056339 | |
M73483.1 | - | 6 | 706 | ||
X65021.1 | - | 6 | 691 |