Gene Transcripts

Gene Symbol
Hao1
Entrez Gene ID
15112
Gene Name
hydroxyacid oxidase 1, liver
Gene Aliases
GOX, Gox1, Hao-1
Chromosome Location
Chr. 2: 134497361 - 134554352 on Build GRCm38
UniGene ID
Mm.26634
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010403.2 NP_034533.1 3, 4 573
GenBank mRNA AF104312.1 - 3, 4 573
AK149565.1 - 3, 4 580
BC018587.1 - 3, 4 551
BC119535.1 - 3, 4 573 40056371
BC119536.1 - 3, 4 573 40056372

View Full Product Details