Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008700.3 | NP_001008700.1 | 4 | 437 | |
GenBank mRNA | AF000304.1 | - | 2 | 214 | |
AK034466.1 | - | 4 | 481 | ||
AK042342.1 | - | 4 | 437 | ||
AK088086.1 | - | 4 | 449 | ||
AK137827.1 | - | 4 | 449 | ||
AK149603.1 | - | 4 | 450 | ||
AK149717.1 | - | 4 | 450 | ||
AK150584.1 | - | 4 | 449 | ||
AK151998.1 | - | 4 | 449 | ||
AK153186.1 | - | 4 | 449 | ||
AK159375.1 | - | 4 | 448 | ||
AK160016.1 | - | 4 | 441 | ||
BC112911.1 | - | 4 | 437 | 30138743 | |
BC128301.1 | - | 3 | 308 | ||
BC128302.1 | - | 3 | 308 | ||
KC212685.1 | - | 329 | |||
M27959.1 | - | 4 | 432 | ||
M27960.1 | - | 4 | 432 | ||
M29854.1 | - | 4 | 416 |