Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146100.1 | NP_001139572.1 | 4 | 776 | |
NM_010438.3 | NP_034568.2 | 7 | 650 | ||
GenBank mRNA | AK143630.1 | - | 4 | 714 | |
AK149749.1 | - | 4 | 489 | ||
AK165237.1 | - | 4 | 577 | ||
AK167468.1 | - | 4 | 532 | ||
BC027316.1 | - | 4 | 487 | ||
BC072628.1 | - | 4 | 897 | 6832742 | |
J05277.1 | - | 4 | 434 | ||
L16949.1 | - | 7 | 629 |