Gene Transcripts

Gene Symbol
Id2
Entrez Gene ID
15902
Gene Name
inhibitor of DNA binding 2
Gene Aliases
AI255428, C78922, Idb2, bHLHb26
Chromosome Location
Chr. 12: 25093799 - 25096092 on Build GRCm38
UniGene ID
Mm.34871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010496.3 NP_034626.1 1, 2 438
GenBank mRNA AK003222.1 - 1, 2 435
AK013239.1 - 1 721
AK013378.1 - 1, 2 435
AK135547.1 - 1, 2 440
AK146346.1 - 1, 2 438
AK146606.1 - 1, 2 441
BC006921.1 - 1, 2 415 2655173
BC053699.1 - 1, 2 425 6515664
M69293.1 - 1, 2 432

View Full Product Details