Gene Transcripts

Gene Symbol
Hoxa5
Entrez Gene ID
15402
Gene Name
homeobox A5
Gene Aliases
Hox-1.3
Chromosome Location
Chr. 6: 52201754 - 52204587 on Build GRCm38
UniGene ID
Mm.173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010453.5 NP_034583.1 2 1802
GenBank mRNA M28021.1 - 2 1800
M36604.1 - 2 1678
X16840.1 - 2 1677

View Full Product Details