Gene Transcripts

Gene Symbol
Hsd17b1
Entrez Gene ID
15485
Gene Name
hydroxysteroid (17-beta) dehydrogenase 1
Gene Aliases
17HSDB1, 17beta-HSD, E2DH, Hsd17ba
Chromosome Location
Chr. 11: 101077968 - 101081188 on Build GRCm38
UniGene ID
Mm.188939
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010475.1 NP_034605.1 6 1299
GenBank mRNA AK143346.1 - 6 1248
X89627.1 - 6 1299

View Full Product Details