Gene Transcripts
- Gene Symbol
- Klra3
- Entrez Gene ID
- 16634
- Gene Name
- killer cell lectin-like receptor, subfamily A, member 3
- Gene Aliases
- 5E6, Ly49c, NK-2.1, Nk-2, Nk2, Nk2.1
- Chromosome Location
- Chr. 6: 130323289 - 130337626 on Build GRCm38
- UniGene ID
- Mm.425338
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289604.1 |
NP_001276533.1
|
2, 3 |
296 |
|
NM_001289605.1 |
NP_001276534.1
|
2, 3 |
330 |
|
NM_010648.3 |
NP_034778.2
|
2, 3 |
296 |
|
GenBank mRNA |
AK159595.1 |
- |
2, 3 |
245 |
|
BC099867.1 |
- |
|
128 |
40040425 |
BC099878.1 |
- |
|
128 |
40040424 |
BC099879.1 |
- |
|
128 |
40040423 |
BC099880.1 |
- |
|
128 |
40040422 |
BC101952.1 |
- |
2, 3 |
172 |
40044699 |
BC103543.1 |
- |
2, 3 |
172 |
40044703 |
BC103544.1 |
- |
2, 3 |
206 |
40044701 |
BC103545.1 |
- |
2, 3 |
172 |
40044700 |
BC132524.1 |
- |
2, 3 |
177 |
40130801 |
BC132526.1 |
- |
2, 3 |
177 |
40130803 |
BC145192.1 |
- |
2, 3 |
177 |
9053743 |
BC145193.1 |
- |
2, 3 |
211 |
9053744 |
U09739.1 |
- |
2, 3 |
296 |
|
U10305.1 |
- |
2, 3 |
212 |
|
U49868.1 |
- |
2, 3 |
277 |
|
U56404.1 |
- |
2, 3 |
171 |
|
U56405.1 |
- |
2, 3 |
171 |
|
View Full Product Details