Gene Transcripts

Gene Symbol
PPM1K
Entrez Gene ID
152926
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1K
Gene Aliases
BDP, MSUDMV, PP2Ckappa, PP2Cm, PTMP, UG0882E07
Chromosome Location
Chr. 4: 88257609 - 88284831 on Build GRCh38
UniGene ID
Hs.43744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152542.4 NP_689755.3 4 1142
XM_006714111.3 XP_006714174.1 4 1203
XM_017007803.1 XP_016863292.1 4 790
XM_017007804.1 XP_016863293.1 5 1330
XM_017007805.1 XP_016863294.1 6 1330
XM_017007806.1 XP_016863295.1 4 663
XM_017007807.1 XP_016863296.1 3 381
GenBank mRNA AF351614.1 - 5 1063
AK054678.1 - 4 870
AK314417.1 - 4 867
AL834271.1 - 4 861
AX766363.1 - 3 657
AY157615.1 - 4 870
AY435431.1 - 4 1047
AY994097.1 - 4 878
BC020850.1 - 4 872
BC037552.1 - 4 906 5227317

View Full Product Details