Gene Transcripts

Gene Symbol
Msx3
Entrez Gene ID
17703
Gene Name
msh homeobox 3
Gene Aliases
AI323377
Chromosome Location
Chr. 7: 140046156 - 140049970 on Build GRCm38
UniGene ID
Mm.4816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010836.3 NP_034966.1 2 1630
GenBank mRNA AK012556.1 - 2 1699
BC051983.1 - 2 1599
U62523.1 - 2 1640

View Full Product Details