Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008542.2 | NP_001008542.2 | 6 | 2125 | |
NM_001008543.2 | NP_001008543.1 | 6 | 1812 | ||
NM_010847.3 | NP_034977.1 | 6 | 1965 | ||
GenBank mRNA | AK140465.1 | - | 6 | 1915 | |
AK144219.1 | - | 6 | 2010 | ||
AK154538.1 | - | 6 | 2115 | ||
BC064453.1 | - | 6 | 2109 | 6810025 | |
BE626657.1 | - | 4 | 453 | ||
U24674.1 | - | 6 | 1744 |