Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010946.3 | NP_035076.1 | 6 | 537 | |
GenBank mRNA | AK041112.1 | - | 6 | 494 | |
AK207258.1 | - | 2 | 85 | ||
AK214245.1 | - | 4 | 285 | ||
BC030172.1 | - | 6 | 535 | 5037501 | |
BY254590.1 | - | 4 | 368 | ||
U57692.1 | - | 6 | 491 |