Gene Transcripts
- Gene Symbol
- Lgals3bp
- Entrez Gene ID
- 19039
- Gene Name
- lectin, galactoside-binding, soluble, 3 binding protein
- Gene Aliases
- 90K, CyCAP, MAC-2BP, Ppicap, Tango10b
- Chromosome Location
- Chr. 11: 118392752 - 118401931 on Build GRCm38
- UniGene ID
- Mm.3152
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011150.2 |
NP_035280.1
|
5, 6 |
805 |
|
GenBank mRNA |
AK079915.1 |
- |
5, 6 |
834 |
|
AK137836.1 |
- |
5, 6 |
782 |
|
AK150021.1 |
- |
5, 6 |
834 |
|
AK152182.1 |
- |
5, 6 |
747 |
|
AK153313.1 |
- |
5, 6 |
821 |
|
AK153396.1 |
- |
5, 6 |
823 |
|
AK155031.1 |
- |
5, 6 |
747 |
|
BC090658.1 |
- |
5, 6 |
801 |
30666981 |
L16894.1 |
- |
5, 6 |
812 |
|
X67809.1 |
- |
5, 6 |
805 |
|
View Full Product Details