Gene Transcripts

Gene Symbol
Psmb7
Entrez Gene ID
19177
Gene Name
proteasome (prosome, macropain) subunit, beta type 7
Gene Aliases
AU020723, MC14
Chromosome Location
Chr. 2: 38588046 - 38643952 on Build GRCm38
UniGene ID
Mm.389251
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011187.1 NP_035317.1 3 208
GenBank mRNA AK002823.1 - 3 223
AK012613.1 - 3 209
AK013961.1 - 3 219
AK031913.1 - 3 224
AK075759.1 - 3 224
AK088276.1 - 3 219
AK088765.1 - 3 222
AK162002.1 - 3 227
AK168823.1 - 3 211
BC033424.1 - 3 205
BC049230.1 - 3 201
BC057662.1 - 3 209 2865594
D83585.1 - 3 208
Y10874.1 - 3 222

View Full Product Details