Gene Transcripts
- Gene Symbol
- PPM1K
- Entrez Gene ID
- 152926
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent 1K
- Gene Aliases
- BDP, MSUDMV, PP2Ckappa, PP2Cm, PTMP, UG0882E07
- Chromosome Location
- Chr. 4: 88257609 - 88284831 on Build GRCh38
- UniGene ID
- Hs.43744
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152542.4 |
NP_689755.3
|
5, 6 |
1338 |
|
XM_006714111.3 |
XP_006714174.1
|
5, 6 |
1399 |
|
XM_017007803.1 |
XP_016863292.1
|
5, 6 |
986 |
|
XM_017007804.1 |
XP_016863293.1
|
6, 7 |
1526 |
|
XM_017007805.1 |
XP_016863294.1
|
7, 8 |
1526 |
|
XM_017007806.1 |
XP_016863295.1
|
5, 6 |
859 |
|
XM_017007807.1 |
XP_016863296.1
|
4, 5 |
577 |
|
GenBank mRNA |
AK054678.1 |
- |
5, 6 |
1066 |
|
AK314417.1 |
- |
5, 6 |
1063 |
|
AL834271.1 |
- |
5, 6 |
1057 |
|
AX766363.1 |
- |
4, 5 |
853 |
|
AY157615.1 |
- |
5, 6 |
1066 |
|
AY435431.1 |
- |
5, 6 |
1243 |
|
AY994097.1 |
- |
5, 6 |
1074 |
|
BC037552.1 |
- |
5, 6 |
1102 |
5227317 |
View Full Product Details