Gene Transcripts

Gene Symbol
NUP214
Entrez Gene ID
8021
Gene Name
nucleoporin 214
Gene Aliases
CAIN, CAN
Chromosome Location
Chr. 9: 131125561 - 131234670 on Build GRCh38
UniGene ID
Hs.654530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318324.1 NP_001305253.1 6 824
NM_005085.3 NP_005076.3 6 824
XM_005272216.3 XP_005272273.1 6 824
XM_005272218.3 XP_005272275.1 6 824
XM_005272219.3 XP_005272276.1 6 824
XM_006717292.3 XP_006717355.1 6 824
XM_011519053.2 XP_011517355.1 6 824
XM_011519055.2 XP_011517357.1 6 824
XM_017015167.1 XP_016870656.1 6 824
GenBank mRNA AB159230.1 - 6 683
AK307573.1 - 6 796
BC012500.1 - 6 768
BC045620.1 - 6 768 5268515
BC105998.1 - 6 799 30531438
D14689.1 - 6 735
X64228.1 - 6 774

View Full Product Details