Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289472.1 | NP_001276401.1 | 5 | 935 | |
NM_011256.3 | NP_035386.1 | 6 | 1126 | ||
GenBank mRNA | AF058693.1 | - | 5 | 794 | |
AK129364.1 | - | 5 | 935 | ||
AK147486.1 | - | 5 | 807 | ||
BC070452.1 | - | 5 | 935 | 30621943 | |
BC082303.1 | - | 6 | 1126 |