Gene Transcripts

Gene Symbol
Slfn1
Entrez Gene ID
20555
Gene Name
schlafen 1
Gene Aliases
AV316259
Chromosome Location
Chr. 11: 83116845 - 83122659 on Build GRCm38
UniGene ID
Mm.10948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011407.1 NP_035537.1 2 378
GenBank mRNA AF099972.1 - 2 378
AK138710.1 - 2 381
BC052869.1 - 2 386 30073520

View Full Product Details