Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163664.1 | NP_001157136.1 | 12 | 330 | |
NM_001163665.1 | NP_001157137.1 | 12 | 330 | ||
NM_001163666.1 | NP_001157138.1 | 10 | 303 | ||
NM_001163667.1 | NP_001157139.1 | 11 | 318 | ||
NM_001163668.1 | NP_001157140.1 | 10 | 279 | ||
NM_001163669.1 | NP_001157141.1 | 11 | 291 | ||
NM_001163670.1 | NP_001157142.1 | 8 | 246 | ||
NM_011620.3 | NP_035750.2 | 9 | 258 | ||
GenBank mRNA | AA028290.1 | - | 11 | 271 | |
AA754928.1 | - | 9 | 234 | ||
AA771596.1 | - | 10 | 256 | ||
AI173874.1 | - | 10 | 295 | ||
AI574395.1 | - | 12 | 324 | ||
AI746973.1 | - | 10 | 298 | ||
AK003147.1 | - | 8 | 213 | ||
BC003747.1 | - | 12 | 326 | 3594256 | |
BY766323.1 | - | 11 | 308 | ||
CB196352.1 | - | 2 | 82 | ||
DV049834.1 | - | 9 | 305 | ||
L48988.1 | - | 6 | 149 | ||
L48989.1 | - | 6 | 149 | ||
L48990.1 | - | 9 | 194 | ||
L48991.1 | - | 6 | 176 | ||
L48992.1 | - | 8 | 182 | ||
L48993.1 | - | 8 | 182 | ||
L49018.1 | - | 6 | 176 | ||
L49466.1 | - | 11 | 318 | ||
L49467.1 | - | 12 | 330 | ||
L49468.1 | - | 10 | 303 | ||
L49469.1 | - | 10 | 303 | ||
L49470.1 | - | 11 | 291 | ||
L49471.1 | - | 10 | 300 | ||
L49472.1 | - | 10 | 279 | ||
U77779.1 | - | 9 | 251 |