Gene Transcripts

Gene Symbol
Tom1
Entrez Gene ID
21968
Gene Name
target of myb1 homolog (chicken)
Gene Aliases
Chromosome Location
Chr. 8: 75033686 - 75070121 on Build GRCm38
UniGene ID
Mm.1967
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136259.1 NP_001129731.1 16 1622
NM_011622.3 NP_035752.1 15 1550
GenBank mRNA AJ006972.1 - 15 1527
AK004063.1 - 5 553
AK028398.1 - 15 1491
AK051245.1 - 15 1532
AK150143.1 - 16 1572
AK160403.1 - 15 1496
AK177998.1 - 3 217
AK181519.1 - 3 243
AK185011.1 - 3 218
AK210394.1 - 3 218
AK214667.1 - 3 248
BC021633.1 - 15 1513 4208226
BC093520.1 - 15 1478 2866013
BC157905.1 - 15 1511 9088106
BC158042.1 - 15 1511 9007343

View Full Product Details