Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127259.1 | NP_001120731.1 | 14 | 3745 | |
NM_001127260.1 | NP_001120732.1 | 13 | 3651 | ||
NM_001127262.1 | NP_001120734.1 | 11 | 2988 | ||
NM_001127264.1 | NP_001120736.1 | 12 | 3070 | ||
NM_011641.2 | NP_035771.1 | 12 | 3082 | ||
GenBank mRNA | AB010152.1 | - | 12 | 3054 | |
AK137253.1 | - | 12 | 3071 | ||
AK137743.1 | - | 1 | 574 | ||
BC090649.1 | - | 12 | 3052 | 30645427 |