Gene Transcripts

Gene Symbol
Zfr
Entrez Gene ID
22763
Gene Name
zinc finger RNA binding protein
Gene Aliases
C920030H05Rik
Chromosome Location
Chr. 15: 12117793 - 12185449 on Build GRCm38
UniGene ID
Mm.273496
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011767.2 NP_035897.2 3, 4 488
GenBank mRNA AF071059.1 - 3, 4 486
BC038599.1 - 3, 4 492
BC058570.1 - 3, 4 488
BC094220.1 - 2, 3 376
BC141139.1 - 3, 4 472 9055751

View Full Product Details