Gene Transcripts

Gene Symbol
Twist1
Entrez Gene ID
22160
Gene Name
twist basic helix-loop-helix transcription factor 1
Gene Aliases
AA960487, M-Twist, Pde, Ska10, Ska<m10Jus>, Twist, bHLHa38, pdt
Chromosome Location
Chr. 12: 33957671 - 33959831 on Build GRCm38
UniGene ID
Mm.3280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011658.2 NP_035788.1 2 1368
GenBank mRNA BC033434.1 - 2 1194
BC083139.1 - 2 1368 6516673
M63650.1 - 2 1227

View Full Product Details