Gene Transcripts

Gene Symbol
Arih2
Entrez Gene ID
23807
Gene Name
ariadne homolog 2 (Drosophila)
Gene Aliases
AI843547, ARI2, TRIAD1, UIP48
Chromosome Location
Chr. 9: 108602942 - 108654719 on Build GRCm38
UniGene ID
Mm.290447
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011790.4 NP_035920.1 3 418
GenBank mRNA AF124664.1 - 2 245
AJ130975.1 - 2 277
AK011094.1 - 3 422
AK045012.1 - 401
AK149936.1 - 3 398
AK165028.1 - 3 356
AK167104.1 - 3 375
AK168055.1 - 3 423
BC051998.1 - 3 418 6400287
BC052422.1 - 3 418 6400287

View Full Product Details