Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011804.2 | NP_035934.1 | 4 | 1913 | |
GenBank mRNA | AK010947.1 | - | 4 | 1929 | |
AK085844.1 | - | 4 | 1726 | ||
AK144101.1 | - | 4 | 1932 | ||
AK150006.1 | - | 5 | 2039 | ||
BC027426.1 | - | 4 | 1913 | 3670649 |