Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012011.1 | NP_036141.1 | 4 | 377 | |
GenBank mRNA | AJ006584.1 | - | 4 | 377 | |
AK009448.1 | - | 4 | 382 | ||
AK153974.1 | - | 4 | 395 | ||
BC031450.1 | - | 4 | 373 | ||
BC043656.1 | - | 4 | 377 | 5368023 |