Gene Transcripts

Gene Symbol
Omd
Entrez Gene ID
27047
Gene Name
osteomodulin
Gene Aliases
OSAD, SLRR2C
Chromosome Location
Chr. 13: 49579754 - 49592612 on Build GRCm38
UniGene ID
Mm.390589
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012050.2 NP_036180.1 3 457
GenBank mRNA AB007848.1 - 3 462
BC095997.1 - 2 263 30739852

View Full Product Details