Gene Transcripts

Gene Symbol
Cpt1a
Entrez Gene ID
12894
Gene Name
carnitine palmitoyltransferase 1a, liver
Gene Aliases
C730027G07, CPTI, Cpt1
Chromosome Location
Chr. 19: 3322326 - 3385735 on Build GRCm38
UniGene ID
Mm.18522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013495.2 NP_038523.2 19 2497
GenBank mRNA AK077454.1 - 19 2451
AK128925.1 - 19 2451
AK133472.1 - 19 2452
AK136487.1 - 19 2451
AK147770.1 - 19 2452
AK149688.1 - 19 2448
BC038395.1 - 18 2432 2649352
BC046383.1 - 18 2432 4160676
BC054791.1 - 19 2478 6832731
D49472.1 - 18 2424
S82796.1 - 2 213

View Full Product Details