Gene Transcripts
- Gene Symbol
- Odf2
- Entrez Gene ID
- 18286
- Gene Name
- outer dense fiber of sperm tails 2
- Gene Aliases
- AI848335, MMTEST29
- Chromosome Location
- Chr. 2: 29889720 - 29931746 on Build GRCm38
- UniGene ID
- Mm.330116
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113213.1 |
NP_001106684.1
|
5, 6 |
752 |
|
NM_001113214.1 |
NP_001106685.1
|
5, 6 |
752 |
|
NM_001177659.1 |
NP_001171130.1
|
5, 6 |
752 |
|
NM_001177661.1 |
NP_001171132.1
|
4, 5 |
598 |
|
NM_013615.3 |
NP_038643.1
|
5, 6 |
725 |
|
GenBank mRNA |
AF000968.1 |
- |
3, 4 |
572 |
|
AF034105.1 |
- |
5, 6 |
688 |
|
AK143726.1 |
- |
5, 6 |
656 |
|
AK148523.1 |
- |
5, 6 |
665 |
|
AK165588.1 |
- |
5, 6 |
656 |
|
AK168415.1 |
- |
5, 6 |
654 |
|
BC057001.1 |
- |
6, 7 |
809 |
6827260 |
BQ840288.1 |
- |
3, 4 |
310 |
|
View Full Product Details