Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163565.1 | NP_001157037.1 | 6 | 903 | |
NM_013643.2 | NP_038671.2 | 6 | 1011 | ||
GenBank mRNA | AK038416.1 | - | 6 | 680 | |
BC051975.1 | - | 6 | 652 | ||
BC057339.1 | - | 5 | 1173 | ||
BC079592.1 | - | 6 | 983 | 6817360 | |
S80329.1 | - | 3 | 847 | ||
U28216.1 | - | 6 | 764 | ||
U28217.1 | - | 6 | 1022 |