Gene Transcripts
- Gene Symbol
- Olfr66
- Entrez Gene ID
- 18367
- Gene Name
- olfactory receptor 66
- Gene Aliases
- 5'[b]1, 5'beta1, MOR1-3, ORL532
- Chromosome Location
- Chr. 7: 103881306 - 103882241 on Build GRCm38
- UniGene ID
- Mm.445781
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013618.3 |
NP_038646.2
|
1 |
698 |
|
- Gene Symbol
- Olfr64
- Entrez Gene ID
- 18366
- Gene Name
- olfactory receptor 64
- Gene Aliases
- 5'[b]2, 5'beta2, MOR1-1, MOR1-2, ORL533
- Chromosome Location
- Chr. 7: 103892810 - 103894471 on Build GRCm38
- UniGene ID
- Mm.377107
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013617.3 |
NP_038645.2
|
1 |
1436 |
|
View Full Product Details