Gene Transcripts

Gene Symbol
Mmel1
Entrez Gene ID
27390
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
Mell1, NEP2, NEPII, Nl1, SEP
Chromosome Location
Chr. 4: 154869585 - 154895530 on Build GRCm38
UniGene ID
Mm.116944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013783.2 NP_038811.2 3 515
GenBank mRNA AF157105.1 - 3 461
AF176569.1 - 3 512
AF302076.1 - 2 205

View Full Product Details