Gene Transcripts

Gene Symbol
Mtf2
Entrez Gene ID
17765
Gene Name
metal response element binding transcription factor 2
Gene Aliases
9230112N11Rik, AA537621, C76717, M96, Pcl2
Chromosome Location
Chr. 5: 108065669 - 108109219 on Build GRCm38
UniGene ID
Mm.257149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253877.1 NP_001240806.1 14 1929
NM_001253878.1 NP_001240807.1 14 1648
NM_001253879.1 NP_001240808.1 13 1571
NM_001253880.1 NP_001240809.1 11 1286
NM_013827.3 NP_038855.2 13 1567
GenBank mRNA AK030752.1 - 13 1517
AK031409.1 - 15 1636
AK032798.1 - 14 1890
AK032819.1 - 14 1891
AK137539.1 - 15 1876
AK139685.1 - 14 1649
BC092237.1 - 13 1499 30637248
BC100340.1 - 11 1217 30715034
S78454.1 - 10 971

View Full Product Details