Gene Transcripts
- Gene Symbol
- Smtn
- Entrez Gene ID
- 29856
- Gene Name
- smoothelin
- Gene Aliases
- smsmo
- Chromosome Location
- Chr. 11: 3517522 - 3539654 on Build GRCm38
- UniGene ID
- Mm.188516
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159284.1 |
NP_001152756.1
|
5, 6 |
582 |
|
NM_001284427.1 |
NP_001271356.1
|
4, 5 |
531 |
|
NM_001284428.1 |
NP_001271357.1
|
5, 6 |
639 |
|
NM_001284429.1 |
NP_001271358.1
|
4, 5 |
531 |
|
NM_013870.3 |
NP_038898.2
|
4, 5 |
531 |
|
GenBank mRNA |
AF064236.1 |
- |
4, 5 |
478 |
|
AJ010305.1 |
- |
4, 5 |
478 |
|
AK030932.1 |
- |
4, 5 |
532 |
|
AK085412.1 |
- |
4, 5 |
493 |
|
AK138898.1 |
- |
4, 5 |
532 |
|
AK166385.1 |
- |
5, 6 |
599 |
|
AK170516.1 |
- |
4, 5 |
491 |
|
BC010599.1 |
- |
4, 5 |
472 |
3709078 |
BC066192.1 |
- |
5, 6 |
460 |
30433616 |
BC069836.1 |
- |
5, 6 |
460 |
30456568 |
BC069839.1 |
- |
5, 6 |
460 |
30431675 |
BY724924.1 |
- |
4, 5 |
483 |
|
CK388087.1 |
- |
1, 2 |
30 |
|
View Full Product Details