Gene Transcripts

Gene Symbol
Neu2
Entrez Gene ID
23956
Gene Name
neuraminidase 2
Gene Aliases
MBS, MSS, MTS
Chromosome Location
Chr. 1: 87574027 - 87597845 on Build GRCm38
UniGene ID
Mm.143717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160163.1 NP_001153635.1 4 575
NM_001160164.1 NP_001153636.1 3 473
NM_001160165.1 NP_001153637.1 3 601
NM_015750.3 NP_056565.1 3 441
GenBank mRNA AB028023.1 - 3 443
AB048604.1 - 3 698
AF139059.1 - 3 552
AK009828.1 - 2 655
AK145284.1 - 3 474
BC026408.1 - 2 229
BC105657.1 - 3 443 40046586
BC105658.1 - 3 443 40046587
BC105659.1 - 3 443 40046588
BC105661.1 - 2 703 40046584

View Full Product Details