Gene Transcripts
- Gene Symbol
- Zpbp
- Entrez Gene ID
- 53604
- Gene Name
- zona pellucida binding protein
- Gene Aliases
- 4930486K01Rik, Iam38, Sp38
- Chromosome Location
- Chr. 11: 11279624 - 11463630 on Build GRCm38
- UniGene ID
- Mm.9500
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001185153.1 |
NP_001172082.1
|
6, 7 |
846 |
|
NM_015785.2 |
NP_056600.1
|
6, 7 |
1017 |
|
GenBank mRNA |
AK006274.1 |
- |
3, 4 |
359 |
|
AK018889.1 |
- |
3, 4 |
361 |
|
AK018921.1 |
- |
3, 4 |
361 |
|
AK029541.1 |
- |
3, 4 |
361 |
|
BC139450.1 |
- |
6, 7 |
986 |
8862472 |
BC139453.1 |
- |
6, 7 |
986 |
8862475 |
BC145628.1 |
- |
6, 7 |
815 |
9054196 |
D17569.1 |
- |
6, 7 |
1006 |
|
View Full Product Details